Assay Details
| Gene Symbol: Ptges | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021583.3 | NP_067594.1 | 1 - 2 | 169 | 105 | |
| GenBank mRNA | AB035145.1 | - | 1 - 2 | 129 | 105 | |
| AB041998.1 | - | 1 - 2 | 158 | 105 | ||
| AB048730.1 | - | 1 - 2 | 171 | 105 | ||
| AF280967.1 | - | 1 - 2 | 168 | 105 | ||
| BC088101.1 | - | 1 - 2 | 127 | 105 | ||
| BC105858.1 | - | 1 - 2 | 155 | 7456259 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptges
Genomic Map