Assay Details
| Gene Symbol: Itgb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147139.2 | NP_671480.2 | 9 - 10 | 1310 | 63 | |
| GenBank mRNA | EV769502.1 | - | 5 - 6 | 641 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itgb5
Genomic Map