Assay Details
| Gene Symbol: Parp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103351.2 | NP_001096821.1 | 4 - 5 | 1184 | 84 | |
| XM_006248411.1 | XP_006248473.1 | 4 - 5 | 1010 | 84 | ||
| XM_008768727.1 | XP_008766949.1 | 4 - 5 | 2293 | 84 | ||
| XM_008768728.1 | XP_008766950.1 | 3 - 4 | 2938 | 84 | ||
| XM_008768729.1 | XP_008766951.1 | 4 - 5 | 1114 | 84 | ||
| GenBank mRNA | BC128756.1 | - | 4 - 5 | 1187 | 7105391 | 84 |
More Information
Updated Mapping Information
Set Membership: |
|

Parp9
Genomic Map