Assay Details
| Gene Symbol: Vcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053864.2 | NP_446316.1 | 11 - 12 | 1570 | 100 | |
| GenBank mRNA | BC060518.1 | - | 11 - 12 | 1570 | 5597843 | 100 |
| U11760.1 | - | 11 - 12 | 1602 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vcp
Genomic Map