Assay Details
| Gene Symbol: Vcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053864.2 | NP_446316.1 | 15 - 16 | 2372 | 65 | |
| GenBank mRNA | BC060518.1 | - | 15 - 16 | 2372 | 5597843 | 65 |
| U11760.1 | - | 15 - 16 | 2404 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair
ubiquitin-dependent protein catabolic process
ER to Golgi vesicle-mediated transport
activation of cysteine-type endopeptidase activity involved in apoptotic process
cellular response to DNA damage stimulus
protein ubiquitination
protein N-linked glycosylation via asparagine
translesion synthesis
ER-associated ubiquitin-dependent protein catabolic process
retrograde protein transport, ER to cytosol
positive regulation of protein complex assembly
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
protein hexamerization
positive regulation of protein catabolic process
protein homooligomerization
aggresome assembly
positive regulation of protein K63-linked deubiquitination
positive regulation of Lys63-specific deubiquitinase activity
receptor binding
protein binding
ATP binding
lipid binding
ATPase activity
protein phosphatase binding
protein domain specific binding
polyubiquitin binding
protein complex binding
deubiquitinase activator activity
identical protein binding
ADP binding
poly(A) RNA binding
ubiquitin-specific protease binding
double-strand break repair
ubiquitin-dependent protein catabolic process
ER to Golgi vesicle-mediated transport
activation of cysteine-type endopeptidase activity involved in apoptotic process
cellular response to DNA damage stimulus
protein ubiquitination
protein N-linked glycosylation via asparagine
translesion synthesis
ER-associated ubiquitin-dependent protein catabolic process
retrograde protein transport, ER to cytosol
positive regulation of protein complex assembly
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
protein hexamerization
positive regulation of protein catabolic process
protein homooligomerization
aggresome assembly
positive regulation of protein K63-linked deubiquitination
positive regulation of Lys63-specific deubiquitinase activity
receptor binding
protein binding
ATP binding
lipid binding
ATPase activity
protein phosphatase binding
protein domain specific binding
polyubiquitin binding
protein complex binding
deubiquitinase activator activity
identical protein binding
ADP binding
poly(A) RNA binding
ubiquitin-specific protease binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P156433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57500 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P156433 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P156433 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P156433 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P156433 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700145748 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77719 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30776 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5402 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700145748 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10876233 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876231 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876232 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876226 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876238 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876229 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876241 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876234 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876235 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876239 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876237 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876236 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876225 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876230 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876228 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876240 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876227 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10876224 | - | - |
| Rat Genome 230 2.0 Array | 1367455_at | - | - |
| Rat Genome 230 2.0 Array | 1367455_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1367455_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1367455_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500019217.rn.1 | TC0500002599.rn.1 | RTA TC |
| Rat Expression Set 230 | 1367455_PM_at | - | - |
| Rat Expression Set 230 | 1367455_at | - | - |
| Rat Expression Set 230 | 1367455_PM_at | - | - |
| Rat Expression Set 230 | 1367455_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10876224 | - | - |
| Rat Genome U34 Set | rc_AI044341_s_at | - | - |
| Rat Genome U34 Set | U11760_at | - | - |
| Rat Genome U34 Set | rc_AI059675_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21583527 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363637 | - | - |
| RatRef-12 Expression BeadChip | ILMN_62115 | - | - |

Vcp
Genomic Map