Assay Details
| Gene Symbol: LOC100909752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749933.3 | XP_003749981.1 | 20 - 21 | 2742 | 128 | |
| Gene Symbol: Col15a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100535.1 | NP_001094005.1 | 2652 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100909752
Genomic Map