Assay Details
| Gene Symbol: Slc35a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107924.1 | NP_001101394.1 | 1 - 2 | 187 | 84 | |
| XM_006237996.2 | XP_006238058.1 | 2 - 3 | 370 | 84 | ||
| GenBank mRNA | FQ213102.1 | - | 2 - 3 | 221 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc35a1
Genomic Map