Assay Details
| Gene Symbol: Faxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271065.1 | NP_001257994.1 | 1 - 2 | 448 | 99 | |
| XM_006237944.2 | XP_006238006.1 | 1 - 2 | 1196 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Faxc
Genomic Map