Assay Details
| Gene Symbol: Rpp25l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106648.2 | NP_001100118.1 | 1 - 2 | 48 | 62 | |
| GenBank mRNA | BC158601.1 | - | 1 - 2 | 48 | 7133091 | 62 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC158601.1, NM_001106648.2 transcripts.
Set Membership: |
|

Rpp25l
Genomic Map