Assay Details
| Gene Symbol: Shb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238148.1 | XP_006238210.1 | 4 - 5 | 1458 | 67 | |
| XM_008763691.1 | XP_008761913.1 | 4 - 5 | 1290 | 67 | ||
More Information
Set Membership: |
|

Shb
Genomic Map