Assay Details
| Gene Symbol: Map7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106270.2 | NP_001099740.2 | 4 - 5 | 645 | 87 | |
| NM_001198638.1 | NP_001185567.1 | 4 - 5 | 421 | 87 | ||
| GenBank mRNA | CV119382.1 | - | 1 - 2 | 96 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Map7
Genomic Map