Assay Details
| Gene Symbol: Heca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107514.1 | NP_001100984.1 | 2 - 3 | 1196 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P398230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30029 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P398230 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P398230 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P398230 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P398230 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102519 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10716987 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10716986 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10716985 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10716984 | - | - |
| Rat Genome 230 2.0 Array | 1379260_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379260_at | - | - |
| Rat Genome 230 2.0 Array | 1383036_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393331_at | - | - |
| Rat Genome 230 2.0 Array | 1383036_at | - | - |
| Rat Genome 230 2.0 Array | 1393331_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393331_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379260_at | - | - |
| Affymetrix HT Rat Focus Array | 1393331_at | - | - |
| Affymetrix HT Rat Focus Array | 1379260_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100032960.rn.1 | TC0100004065.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100032962.rn.1 | TC0100004065.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100032961.rn.1 | TC0100004065.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393331_PM_at | - | - |
| Rat Expression Set 230 | 1379260_PM_at | - | - |
| Rat Expression Set 230 | 1379260_at | - | - |
| Rat Expression Set 230 | 1393331_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10716984 | - | - |
| Rat Genome U34 Set | rc_AI178526_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20721705 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350409 | - | - |

Heca
Genomic Map