Assay Details
| Gene Symbol: RGD1560020_predicted | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008758639.1 | XP_008756861.1 | 10 - 11 | 3960 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1560020_predicted| Gene Symbol: RGD1560020_predicted | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008758639.1 | XP_008756861.1 | 10 - 11 | 3960 | 113 | |
Set Membership: |
|