Assay Details
| Gene Symbol: Popdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037369.1 | NP_001032446.1 | 4 - 5 | 821 | 102 | |
| XM_006256599.2 | XP_006256661.1 | 3 - 4 | 1061 | 102 | ||
| GenBank mRNA | AY787642.1 | - | 2 - 3 | 788 | 102 | |
| BC105761.1 | - | 4 - 5 | 821 | 5623862 | 102 | |
| FQ218039.1 | - | 4 - 5 | 724 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Popdc3
Genomic Map