Assay Details
| Gene Symbol: Sumo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009672.1 | NP_001009672.1 | 3 - 4 | 208 | 87 | |
| GenBank mRNA | BC088322.1 | - | 3 - 4 | 208 | 7325996 | 87 |
| FQ211636.1 | - | 3 - 4 | 245 | 87 | ||
| FQ214927.1 | - | 3 - 4 | 278 | 87 | ||
| FQ215001.1 | - | 3 - 4 | 288 | 87 | ||
| FQ215643.1 | - | 3 - 4 | 253 | 87 | ||
| FQ224870.1 | - | 3 - 4 | 277 | 87 | ||
| FQ234204.1 | - | 3 - 4 | 280 | 87 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sumo1
Genomic Map