Assay Details
| Gene Symbol: LOC100911373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750700.3 | XP_003750748.1 | 1 - 2 | 192 | 105 | |
| XM_006244941.2 | XP_006245003.1 | 1 - 2 | 193 | 105 | ||
| XM_008767154.1 | XP_008765376.1 | 1 - 2 | 194 | 105 | ||
| XM_008767155.1 | XP_008765377.1 | 1 - 2 | 193 | 105 | ||
| Gene Symbol: Boll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113370.1 | NP_001106841.1 | 1 - 2 | 183 | 105 | |
| XM_006244929.2 | XP_006244991.1 | 1 - 2 | 193 | 105 | ||
| XM_008767125.1 | XP_008765347.1 | 1 - 2 | 194 | 105 | ||
| XM_008767126.1 | XP_008765348.1 | 1 - 2 | 192 | 105 | ||
| XM_008767127.1 | XP_008765349.1 | 1 - 2 | 193 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001113370.1, XM_003750700.3, XM_006244929.2, XM_006244941.2, XM_008767125.1, XM_008767126.1, XM_008767127.1, XM_008767154.1, XM_008767155.1 transcripts.
Set Membership: |
|

LOC100911373
Genomic Map