Assay Details
| Gene Symbol: LOC100911373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750700.3 | XP_003750748.1 | 8 - 9 | 810 | 120 | |
| XM_006244941.2 | XP_006245003.1 | 8 - 9 | 811 | 120 | ||
| XM_008767154.1 | XP_008765376.1 | 8 - 9 | 812 | 120 | ||
| XM_008767155.1 | XP_008765377.1 | 8 - 9 | 811 | 120 | ||
| Gene Symbol: Boll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113370.1 | NP_001106841.1 | 8 - 9 | 801 | 120 | |
| XM_006244929.2 | XP_006244991.1 | 8 - 9 | 811 | 120 | ||
| XM_008767125.1 | XP_008765347.1 | 8 - 9 | 812 | 120 | ||
| XM_008767126.1 | XP_008765348.1 | 8 - 9 | 810 | 120 | ||
| XM_008767127.1 | XP_008765349.1 | 8 - 9 | 811 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100911373
Genomic Map