Assay Details
| Gene Symbol: Crygd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033095.1 | NP_149086.1 | 2 - 3 | 295 | 57 | |
| XM_008767074.1 | XP_008765296.1 | 1 - 2 | 1166 | 57 | ||
| GenBank mRNA | X57169.1 | - | 2 - 3 | 280 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crygd
Genomic Map