Assay Details
| Gene Symbol: Hs6st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108210.1 | NP_001101680.1 | 1 - 2 | 341 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P046353 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P145970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157629 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P046353 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P145970 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P046353 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P145970 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P046353 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P145970 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P20311 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10922490 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922491 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10922489 | - | - |
| Rat Genome 230 2.0 Array | 1372467_at | - | - |
| Rat Genome 230 2.0 Array | 1372467_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372467_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372467_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900003626.rn.1 | TC0900000469.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900003627.rn.1 | TC0900000469.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900003625.rn.1 | TC0900000469.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372467_PM_at | - | - |
| Rat Expression Set 230 | 1372467_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922489 | - | - |
| Rat Genome U34 Set | rc_AA943794_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21484332 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358565 | - | - |

Hs6st1
Genomic Map