Assay Details
| Gene Symbol: Eif4e1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771512.1 | XP_008769734.1 | 7 - 8 | 626 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif4e1b| Gene Symbol: Eif4e1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771512.1 | XP_008769734.1 | 7 - 8 | 626 | 54 | |
Set Membership: |
|