Assay Details
Gene Symbol: Dok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107336.1 | NP_001100806.1 | 3 - 4 | 491 | 72 | |
XM_006253619.2 | XP_006253681.1 | 3 - 4 | 503 | 72 | ||
GenBank mRNA | BC166772.1 | - | 3 - 4 | 483 | 7934143 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
