Assay Details
| Gene Symbol: Slc4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017048.2 | NP_058744.1 | 5 - 6 | 775 | 78 | |
| XM_006235874.2 | XP_006235936.1 | 5 - 6 | 1232 | 78 | ||
| XM_006235875.2 | XP_006235937.1 | 5 - 6 | 953 | 78 | ||
| XM_006235876.2 | XP_006235938.1 | 5 - 6 | 1015 | 78 | ||
| XM_006235877.2 | XP_006235939.1 | 4 - 5 | 1158 | 78 | ||
| XM_006235878.2 | XP_006235940.1 | 4 - 5 | 1077 | 78 | ||
| GenBank mRNA | AJ288902.1 | - | 4 - 5 | 570 | 78 | |
| J05166.1 | - | 5 - 6 | 779 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a2
Genomic Map