Assay Details
| Gene Symbol: LOC499971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024317.2 | NP_001019488.2 | 17 - 18 | 2148 | 92 | |
| XM_008762655.1 | XP_008760877.1 | 16 - 17 | 1801 | 92 | ||
| XM_008762656.1 | XP_008760878.1 | 15 - 16 | 1692 | 92 | ||
| GenBank mRNA | BC079392.1 | - | 10 - 11 | 1084 | 7111961 | 92 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC499971
Genomic Map