Assay Details
| Gene Symbol: Gnat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173139.1 | NP_775162.1 | 2 - 3 | 277 | 128 | |
| GenBank mRNA | X65747.1 | - | 2 - 3 | 277 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnat3
Genomic Map