Assay Details
| Gene Symbol: Atg9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191968.1 | NP_001178897.1 | 7 - 8 | 1881 | 61 | |
| XM_006235960.2 | XP_006236022.1 | 7 - 8 | 1881 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atg9b
Genomic Map