Assay Details
| Gene Symbol: LOC100909675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110709.1 | 1 | 221 | 85 | ||
| GenBank mRNA | FN803941.1 | - | 1 | 221 | 85 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100909675
Genomic Map