Assay Details
| Gene Symbol: Sema3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106578.1 | NP_001100048.1 | 5 - 6 | 551 | 82 | |
| XM_008762645.1 | XP_008760867.1 | 6 - 7 | 860 | 82 | ||
| XM_008762646.1 | XP_008760868.1 | 6 - 7 | 858 | 82 | ||
| GenBank mRNA | BC169108.1 | - | 5 - 6 | 551 | 7096896 | 82 |
More Information
Updated Mapping Information
Set Membership: |
|

Sema3c
Genomic Map