Assay Details
| Gene Symbol: Orc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014186.1 | NP_001014208.1 | 9 - 10 | 958 | 119 | |
| XM_006235947.2 | XP_006236009.1 | 9 - 10 | 1014 | 119 | ||
| GenBank mRNA | BC088447.1 | - | 9 - 10 | 958 | 7126516 | 119 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P131012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73441 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P131012 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P131012 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P131012 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17671 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P131012 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73444 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99000 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 87016 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49569 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21078 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10853018 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853009 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853006 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853007 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853012 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853008 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853016 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853013 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853015 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853017 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853011 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853005 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853019 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853010 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10853004 | - | - |
| Rat Genome 230 2.0 Array | 1373711_at | - | - |
| Rat Genome 230 2.0 Array | 1374264_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374264_at | - | - |
| Rat Genome 230 2.0 Array | 1373711_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373711_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374264_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374264_at | - | - |
| Affymetrix HT Rat Focus Array | 1373711_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400001047.rn.1 | TC0400000144.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400001050.rn.1 | TC0400000144.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373711_PM_at | - | - |
| Rat Expression Set 230 | 1374264_at | - | - |
| Rat Expression Set 230 | 1373711_at | - | - |
| Rat Expression Set 230 | 1374264_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853004 | - | - |
| Rat Genome U34 Set | rc_AI234664_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21715637 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356597 | - | - |

Orc5
Genomic Map