Assay Details
| Gene Symbol: NEWGENE_2301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EF116601.1 | - | 1 - 2 | 72 | 112 | |
| Gene Symbol: Cd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031561.2 | NP_113749.2 | 39 | 112 | ||
| GenBank mRNA | BC072543.1 | - | 39 | 7105943 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC072543.1, NM_031561.2 transcripts.
Set Membership: |
|

NEWGENE_2301
Genomic Map