Assay Details
| Gene Symbol: Pcmtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763511.1 | XP_008761733.1 | 2 - 3 | 1153 | 123 | |
| XM_008763513.1 | XP_008761735.1 | 2 - 3 | 1153 | 123 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008763511.1, XM_008763513.1 transcripts.
Set Membership: |
|

Pcmtd1
Genomic Map