Assay Details
| Gene Symbol: Gal3st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006251383.2 | XP_006251445.1 | 1 - 2 | 1934 | 73 | |
| GenBank mRNA | BC087145.1 | - | 1 - 2 | 70 | 7129361 | 73 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC087145.1, XM_006251383.2 transcripts.
Set Membership: |
|

Gal3st1
Genomic Map