Assay Details
| Gene Symbol: Nsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024128.3 | NP_077042.2 | 1 - 2 | 52 | 68 | |
| GenBank mRNA | BC060538.1 | - | 1 - 2 | 55 | 6921928 | 68 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC060538.1, NM_024128.3 transcripts.
Set Membership: |
|

Nsg1
Genomic Map