Assay Details
| Gene Symbol: Nsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024128.3 | NP_077042.2 | 2 - 3 | 212 | 74 | |
| GenBank mRNA | BC060538.1 | - | 2 - 3 | 215 | 6921928 | 74 |
More Information
Updated Mapping Information
Set Membership: |
|

Nsg1
Genomic Map