Assay Details
| Gene Symbol: Lif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006251364.2 | XP_006251426.1 | 2 - 3 | 150 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006251364.2 transcripts.
Set Membership: |
|

Lif
Genomic Map