Assay Details
| Gene Symbol: Osbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107232.1 | NP_001100702.1 | 3 | 810 | 69 | |
| XM_008770297.1 | XP_008768519.1 | 3 | 843 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Osbp2
Genomic Map