Assay Details
| Gene Symbol: Rab28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053978.1 | NP_446430.1 | 5 - 6 | 513 | 72 | |
| XM_008770195.1 | XP_008768417.1 | 5 - 6 | 882 | 72 | ||
| XM_008770196.1 | XP_008768418.1 | 5 - 6 | 882 | 72 | ||
| XM_008770197.1 | XP_008768419.1 | 5 - 6 | 1091 | 72 | ||
| XM_008770198.1 | XP_008768420.1 | 5 - 6 | 882 | 72 | ||
| GenBank mRNA | BC072512.1 | - | 5 - 6 | 637 | 7104824 | 72 |
| X78606.1 | - | 5 - 6 | 513 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab28
Genomic Map