Assay Details
| Gene Symbol: Clnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006251106.2 | XP_006251168.1 | 6 - 7 | 566 | 64 | |
| XM_006251108.2 | XP_006251170.1 | 6 - 7 | 566 | 64 | ||
| XM_008770228.1 | XP_008768450.1 | 3 - 4 | 412 | 64 | ||
| XM_008770229.1 | XP_008768451.1 | 3 - 4 | 412 | 64 | ||
| XM_008770230.1 | XP_008768452.1 | 3 - 4 | 412 | 64 | ||
More Information
Set Membership: |
|

Clnk
Genomic Map