Assay Details
| Gene Symbol: Fam175a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009633.1 | NP_001009633.1 | 2 - 3 | 227 | 143 | |
| GenBank mRNA | BC088408.1 | - | 2 - 3 | 227 | 7131771 | 143 |
More Information
Updated Mapping Information
Set Membership: |
|

Fam175a
Genomic Map