Assay Details
| Gene Symbol: Nxpe3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109435.1 | NP_001102905.1 | 4 - 5 | 1411 | 112 | |
| XM_006248252.2 | XP_006248314.1 | 4 - 5 | 1237 | 112 | ||
| XM_006248253.2 | XP_006248315.1 | 4 - 5 | 1248 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nxpe3
Genomic Map