Assay Details
| Gene Symbol: Trat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037981.1 | NP_001033070.1 | 4 - 5 | 412 | 119 | |
| GenBank mRNA | BC111081.1 | - | 4 - 5 | 412 | 7384735 | 119 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P886413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37219 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P886413 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P886413 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P886413 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P886413 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96246 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 32351 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100100 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 7735 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59273 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10750959 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750964 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750962 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750960 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750965 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750961 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750963 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10750958 | - | - |
| Rat Genome 230 2.0 Array | 1393791_at | - | - |
| Rat Genome 230 2.0 Array | 1393791_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100002812.rn.1 | TC1100000548.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100002811.rn.1 | TC1100000548.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393791_at | - | - |
| Rat Expression Set 230 | 1393791_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750958 | - | - |
| Rat Genome U34 Set | rc_AI044622_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21115905 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650313 | - | - |

Trat1
Genomic Map