Assay Details
| Gene Symbol: Zfp148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031615.1 | NP_113803.1 | 6 - 7 | 964 | 103 | |
| XM_006248435.2 | XP_006248497.1 | 3 - 4 | 612 | 103 | ||
| GenBank mRNA | U30381.1 | - | 6 - 7 | 964 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp148
Genomic Map