Assay Details
| Gene Symbol: Tomm70a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212519.2 | NP_997684.1 | 1 - 2 | 469 | 60 | |
| XM_008768643.1 | XP_008766865.1 | 1 - 2 | 763 | 60 | ||
| XM_008768644.1 | XP_008766866.1 | 1 - 2 | 769 | 60 | ||
| XM_008768645.1 | XP_008766867.1 | 1 - 2 | 767 | 60 | ||
| GenBank mRNA | AB162856.1 | - | 1 - 2 | 332 | 60 | |
| BC098640.1 | - | 1 - 2 | 469 | 7099109 | 60 | |
| FQ228582.1 | - | 513 | 60 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tomm70a
Genomic Map