Assay Details
| Gene Symbol: Fam178a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134612.1 | NP_001128084.1 | 14 - 15 | 3107 | 110 | |
| XM_006231550.2 | XP_006231612.1 | 13 - 14 | 3099 | 110 | ||
| XM_006231551.2 | XP_006231613.1 | 13 - 14 | 3095 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam178a
Genomic Map