Assay Details
| Gene Symbol: Fam178a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134612.1 | NP_001128084.1 | 18 - 19 | 3521 | 162 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P185958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809515 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96414 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P547107 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P547107 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P547107 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1029654 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10715613 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715607 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715605 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715606 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715601 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715603 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715610 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715615 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715604 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715608 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715617 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715612 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715602 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715609 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715600 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715611 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715614 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10715598 | - | - |
| Rat Genome 230 2.0 Array | 1391003_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391003_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100030017.rn.1 | TC0100003691.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100030016.rn.1 | TC0100003691.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100030018.rn.1 | TC0100003691.rn.1 | RTA TC |
| Rat Expression Set 230 | 1391003_PM_at | - | - |
| Rat Expression Set 230 | 1391003_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715598 | - | - |
| Rat Genome U34 Set | rc_AA926245_at | - | - |
| Rat Genome U34 Set | rc_AI012092_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21364092 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368135 | - | - |

Fam178a
Genomic Map