Assay Details
| Gene Symbol: Fam178a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134612.1 | NP_001128084.1 | 7 - 8 | 2203 | 74 | |
| XM_006231550.2 | XP_006231612.1 | 6 - 7 | 2192 | 74 | ||
| XM_006231551.2 | XP_006231613.1 | 6 - 7 | 2191 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam178a
Genomic Map