Assay Details
| Gene Symbol: Fam204a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107448.2 | NP_001100918.1 | 5 - 6 | 622 | 109 | |
| XM_006231673.2 | XP_006231735.1 | 5 - 6 | 1438 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam204a
Genomic Map