Assay Details
| Gene Symbol: Avpi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134373.2 | NP_599200.2 | 2 - 3 | 515 | 67 | |
| XM_006231383.2 | XP_006231445.1 | 2 - 3 | 581 | 67 | ||
| GenBank mRNA | AJ294544.1 | - | 2 - 3 | 67 | ||
| FQ220140.1 | - | 2 - 3 | 425 | 67 | ||
| FQ220957.1 | - | 2 - 3 | 399 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Avpi1
Genomic Map