Assay Details
| Gene Symbol: Rab11fip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107447.2 | NP_001100917.2 | 3 - 4 | 1263 | 71 | |
| GenBank mRNA | BC098050.1 | - | 1301 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab11fip2
Genomic Map