Assay Details
| Gene Symbol: Mapk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031622.2 | NP_113810.2 | 4 - 5 | 1626 | 77 | |
| XM_006243409.2 | XP_006243471.1 | 4 - 5 | 2120 | 77 | ||
| GenBank mRNA | BC109380.1 | - | 4 - 5 | 1577 | 7446401 | 77 |
| M64301.1 | - | 3 - 4 | 1254 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk6
Genomic Map