Assay Details
| Gene Symbol: Tbx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108173.1 | NP_001101643.1 | 7 - 8 | 1537 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
somitogenesis
positive regulation of mesenchymal cell proliferation
sinoatrial node development
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
sensory perception of sound
biological_process
anterior/posterior axis specification
morphogenesis of embryonic epithelium
positive regulation of cell-cell adhesion
cell differentiation
anatomical structure formation involved in morphogenesis
positive regulation of epithelial cell proliferation
smooth muscle cell differentiation
negative regulation of canonical Wnt signaling pathway involved in neural plate anterior/posterior pattern formation
ureter development
cochlea morphogenesis
positive regulation of mesenchymal cell proliferation involved in ureter development
transcription regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II transcription corepressor activity
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
protein homodimerization activity
protein heterodimerization activity
negative regulation of transcription from RNA polymerase II promoter
somitogenesis
positive regulation of mesenchymal cell proliferation
sinoatrial node development
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
sensory perception of sound
biological_process
anterior/posterior axis specification
morphogenesis of embryonic epithelium
positive regulation of cell-cell adhesion
cell differentiation
anatomical structure formation involved in morphogenesis
positive regulation of epithelial cell proliferation
smooth muscle cell differentiation
negative regulation of canonical Wnt signaling pathway involved in neural plate anterior/posterior pattern formation
ureter development
cochlea morphogenesis
positive regulation of mesenchymal cell proliferation involved in ureter development
transcription regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II transcription corepressor activity
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
protein homodimerization activity
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P378639 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P026900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79748 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026900 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P378639 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P378639 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026900 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P026900 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P378639 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P445895 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P559462 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10919179 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919178 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919182 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919180 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919177 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919183 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919181 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919176 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10919175 | - | - |
| Rat Genome 230 2.0 Array | 1379883_at | - | - |
| Rat Genome 230 2.0 Array | 1391630_at | - | - |
| Rat Genome 230 2.0 Array | 1379883_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391630_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391630_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391630_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800024675.rn.1 | TC0800003071.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800024677.rn.1 | TC0800003071.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800024678.rn.1 | TC0800003071.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800024679.rn.1 | TC0800003071.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800024674.rn.1 | TC0800003071.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800024673.rn.1 | TC0800003071.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800024682.rn.1 | TC0800003071.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800024680.rn.1 | TC0800003071.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800024681.rn.1 | TC0800003071.rn.1 | RTA TC |
| Rat Expression Set 230 | 1391630_PM_at | - | - |
| Rat Expression Set 230 | 1391630_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919175 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21990298 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650771 | - | - |

Tbx18
Genomic Map